Assay Details
| Gene Symbol: COX5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007517.1 | NP_001007518.1 | 4 | 349 | 103 | |
| GenBank mRNA | AK343626.1 | - | 4 | 365 | 103 | |
| AK344883.1 | - | 4 | 351 | 103 | ||
| AK392358.1 | - | 4 | 354 | 103 | ||
| AK400056.1 | - | 4 | 423 | 103 | ||
| AY786556.1 | - | 4 | 349 | 103 | ||
More Information
Set Membership: |
|

COX5B
Genomic Map