Assay Details
| Gene Symbol: SAR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008689.2 | NP_001008689.1 | 6 | 548 | 85 | |
| GenBank mRNA | AK232347.1 | - | 6 | 546 | 85 | |
| AK393054.1 | - | 4 | 348 | 85 | ||
| AK396234.1 | - | 6 | 548 | 85 | ||
| AY609439.1 | - | 6 | 491 | 85 | ||
| AY819557.2 | - | 6 | 548 | 85 | ||
| DQ002890.1 | - | 5 | 409 | 85 | ||
| EF190235.1 | - | 5 | 409 | 85 | ||
More Information
Set Membership: |
|

SAR1B
Genomic Map