Assay Details
| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025218.2 | NP_001020389.1 | 4 | 251 | 83 | |
| GenBank mRNA | AY633618.1 | - | 4 | 251 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5MC1| Gene Symbol: ATP5MC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025218.2 | NP_001020389.1 | 4 | 251 | 83 | |
| GenBank mRNA | AY633618.1 | - | 4 | 251 | 83 | |
Set Membership: |
|