Assay Details
| Gene Symbol: MAPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025224.2 | NP_001020395.1 | 12 | 1045 | 81 | |
| GenBank mRNA | AK394752.1 | - | 3 | 263 | 81 | |
| DQ058292.2 | - | 12 | 1045 | 81 | ||
More Information
Set Membership: |
|

MAPK12
Genomic Map