Assay Details
| Gene Symbol: ATP6V0E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044579.1 | NP_001038044.1 | 1 | 141 | 86 | |
| XM_005657621.3 | XP_005657678.1 | 181 | 86 | |||
| GenBank mRNA | AK236159.1 | - | 1 | 115 | 86 | |
| AK237166.1 | - | 1 | 115 | 86 | ||
| AK392853.1 | - | 1 | 142 | 86 | ||
| AY633619.1 | - | 1 | 141 | 86 | ||
More Information
Set Membership: |
|

ATP6V0E1
Genomic Map