Assay Details
| Gene Symbol: RGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044600.1 | NP_001038065.1 | 5 | 554 | 102 | |
| GenBank mRNA | AB530146.1 | - | 5 | 538 | 102 | |
| AK240365.1 | - | 5 | 568 | 102 | ||
| AY609617.1 | - | 5 | 559 | 102 | ||
| DQ150111.2 | - | 5 | 554 | 102 | ||
More Information
Set Membership: |
|

RGS2
Genomic Map