Assay Details
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048071.1 | NP_001041536.1 | 5 | 548 | 63 | |
| GenBank mRNA | AY864609.1 | - | 5 | 548 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NIP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048071.1 | NP_001041536.1 | 5 | 548 | 63 | |
| GenBank mRNA | AY864609.1 | - | 5 | 548 | 63 | |
Set Membership: |
|