Assay Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213743.1 | NP_998908.1 | 2 | 253 | 60 | |
| GenBank mRNA | AK235806.1 | - | 2 | 160 | 60 | |
| AK346933.1 | - | 2 | 160 | 60 | ||
| AK390424.1 | - | 2 | 234 | 60 | ||
| AY234859.1 | - | 2 | 253 | 60 | ||
| AY610356.1 | - | 2 | 253 | 60 | ||
More Information
Set Membership: |
|

FUNDC2
Genomic Map