Assay Details
| Gene Symbol: PAEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213754.2 | NP_998919.1 | 523 | 93 | ||
| GenBank mRNA | AY372187.1 | - | 516 | 93 | ||
| X54976.1 | - | 556 | 93 | |||
More Information
Set Membership: |
|

PAEP
Genomic Map