Assay Details
| Gene Symbol: RSC1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213793.1 | NP_998958.1 | 1941 | 96 | ||
| GenBank mRNA | AK389781.1 | - | 1 | 1312 | 96 | |
| X64315.2 | - | 1941 | 96 | |||
More Information
Set Membership: |
|

RSC1A1
Genomic Map