Assay Details
Gene Symbol: OBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213796.1 | NP_998961.1 | 5 | 447 | 114 | |
GenBank mRNA | AF436848.1 | - | 5 | 447 | 114 |
Gene Symbol: LOC100626157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021080877.1 | XP_020936536.1 | 459 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
