Assay Details
| Gene Symbol: DYNLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195114.2 | NP_001182043.1 | 3 | 278 | 66 | |
| GenBank mRNA | AF436777.2 | - | 2 | 150 | 66 | |
| AK233745.1 | - | 3 | 252 | 66 | ||
| AK236341.1 | - | 3 | 278 | 66 | ||
| AK346797.1 | - | 3 | 246 | 66 | ||
| AK346879.1 | - | 3 | 246 | 66 | ||
| AK349091.1 | - | 3 | 250 | 66 | ||
| AK397363.1 | - | 3 | 246 | 66 | ||
| FN401365.1 | - | 3 | 193 | 66 | ||
More Information
Set Membership: |
|

DYNLL1
Genomic Map