Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213834.1 | NP_998999.1 | 3 | 1045 | 66 | |
| GenBank mRNA | U27083.1 | - | 3 | 1045 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213834.1 | NP_998999.1 | 3 | 1045 | 66 | |
| GenBank mRNA | U27083.1 | - | 3 | 1045 | 66 | |
Set Membership: |
|