Assay Details
Gene Symbol: FCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213868.1 | NP_999033.1 | 1 | 56 | 97 | |
GenBank mRNA | AK231195.1 | - | 1 | 314 | 97 | |
AK231454.1 | - | 1 | 63 | 97 | ||
AK231561.1 | - | 1 | 129 | 97 | ||
AK231689.1 | - | 1 | 69 | 97 | ||
AK232513.1 | - | 1 | 63 | 97 | ||
AK345640.1 | - | 1 | 63 | 97 | ||
L12344.1 | - | 1 | 56 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
