Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214082.1 | NP_999247.1 | 4 | 345 | 64 | |
| GenBank mRNA | AB090164.1 | - | 4 | 345 | 64 | |
| AY609864.1 | - | 4 | 267 | 64 | ||
More Information
Set Membership: |
|

RAMP2
Genomic Map