Assay Details
| Gene Symbol: AKAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214138.1 | NP_999303.1 | 13 - 14 | 1846 | 64 | |
| GenBank mRNA | AF208387.1 | - | 13 - 14 | 1846 | 64 | |
More Information
Set Membership: |
|

AKAP10
Genomic Map