Assay Details
| Gene Symbol: FCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214160.2 | NP_999325.1 | 8 - 9 | 743 | 64 | |
| XM_013988963.2 | XP_013844417.1 | 678 | 64 | |||
| XM_021082862.1 | XP_020938521.1 | 1241 | 64 | |||
| GenBank mRNA | AK232015.1 | - | 8 - 9 | 765 | 64 | |
| AK348880.1 | - | 8 - 9 | 761 | 64 | ||
| L12345.1 | - | 8 - 9 | 743 | 64 | ||
More Information
Set Membership: |
|

FCN1
Genomic Map