Assay Details
| Gene Symbol: DHDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214166.1 | NP_999331.1 | 7 | 952 | 77 | |
| GenBank mRNA | AB021929.1 | - | 7 | 952 | 77 | |
| AK231806.1 | - | 7 | 1012 | 77 | ||
| AK398155.1 | - | 7 | 1036 | 77 | ||
| AK400980.1 | - | 7 | 1036 | 77 | ||
More Information
Set Membership: |
|

DHDH
Genomic Map