Assay Details
| Gene Symbol: UBA52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214211.1 | NP_999376.1 | 5 | 346 | 87 | |
| XM_005652916.3 | XP_005652973.1 | 567 | 87 | |||
| XM_013991270.1 | XP_013846724.1 | 382 | 87 | |||
| XM_013991276.2 | XP_013846730.1 | 610 | 87 | |||
| GenBank mRNA | AK231236.1 | - | 5 | 349 | 87 | |
| AK233792.1 | - | 5 | 352 | 87 | ||
| AK392699.1 | - | 5 | 369 | 87 | ||
| AK392700.1 | - | 5 | 353 | 87 | ||
| AK399521.1 | - | 5 | 364 | 87 | ||
| U72496.1 | - | 5 | 346 | 87 | ||
More Information
Set Membership: |
|

UBA52
Genomic Map