Assay Details
| Gene Symbol: UOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214270.1 | NP_999435.1 | 7 - 8 | 848 | 105 | |
| GenBank mRNA | AK346310.1 | - | 7 - 8 | 888 | 105 | |
| M27697.1 | - | 7 - 8 | 848 | 105 | ||
More Information
Set Membership: |
|

UOX
Genomic Map