Assay Details
| Gene Symbol: CDKN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214316.1 | NP_999481.1 | 2 | 553 | 87 | |
| GenBank mRNA | AB031955.1 | - | 2 | 553 | 87 | |
| AB031956.1 | - | 2 | 553 | 87 | ||
| AB031957.1 | - | 2 | 553 | 87 | ||
More Information
Set Membership: |
|

CDKN1B
Genomic Map