Assay Details
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214332.1 | NP_999497.1 | 6 | 851 | 56 | |
GenBank mRNA | AK237420.1 | - | 6 | 867 | 56 | |
AK237743.1 | - | 6 | 867 | 56 | ||
AK239515.1 | - | 6 | 869 | 56 | ||
AK239567.1 | - | 6 | 868 | 56 | ||
AK345704.1 | - | 6 | 867 | 56 | ||
AK347796.1 | - | 6 | 865 | 56 | ||
AK351147.1 | - | 6 | 868 | 56 | ||
AK392619.1 | - | 6 | 870 | 56 | ||
AK393801.1 | - | 6 | 867 | 56 | ||
AK393848.1 | - | 6 | 871 | 56 | ||
AK393922.1 | - | 6 | 871 | 56 | ||
AK394803.1 | - | 6 | 896 | 56 | ||
AK395551.1 | - | 6 | 897 | 56 | ||
AK396080.1 | - | 6 | 867 | 56 | ||
AK396286.1 | - | 5 | 1385 | 56 | ||
AK396893.1 | - | 6 | 867 | 56 | ||
AK397800.1 | - | 6 | 1019 | 56 | ||
AK398072.1 | - | 6 | 866 | 56 | ||
AK398128.1 | - | 6 | 867 | 56 | ||
AK398252.1 | - | 6 | 867 | 56 | ||
AK398290.1 | - | 6 | 867 | 56 | ||
AK398315.1 | - | 6 | 868 | 56 | ||
AK398526.1 | - | 6 | 868 | 56 | ||
AK398648.1 | - | 6 | 867 | 56 | ||
AK398658.1 | - | 6 | 867 | 56 | ||
AK398662.1 | - | 6 | 867 | 56 | ||
AK399003.1 | - | 6 | 867 | 56 | ||
AK399124.1 | - | 6 | 897 | 56 | ||
AK399358.1 | - | 6 | 898 | 56 | ||
AK399402.1 | - | 6 | 867 | 56 | ||
AK399404.1 | - | 6 | 867 | 56 | ||
AK399406.1 | - | 6 | 867 | 56 | ||
AK399977.1 | - | 5 | 1427 | 56 | ||
AK400504.1 | - | 6 | 896 | 56 | ||
AK400828.1 | - | 6 | 866 | 56 | ||
AK400932.1 | - | 6 | 868 | 56 | ||
AK400960.1 | - | 6 | 898 | 56 | ||
AK400972.1 | - | 6 | 868 | 56 | ||
AK400995.1 | - | 6 | 867 | 56 | ||
AK401007.1 | - | 6 | 867 | 56 | ||
AK401013.1 | - | 6 | 867 | 56 | ||
AK401019.1 | - | 6 | 867 | 56 | ||
AK401023.1 | - | 6 | 870 | 56 | ||
Z33879.1 | - | 851 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
