Assay Details
Gene Symbol: B3GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214351.1 | NP_999516.1 | 1 | 152 | 65 | |
XM_005652413.3 | XP_005652470.1 | 1277 | 65 | |||
XM_021068551.1 | XP_020924210.1 | 1359 | 65 | |||
XM_021068552.1 | XP_020924211.1 | 1418 | 65 | |||
XM_021068553.1 | XP_020924212.1 | 1292 | 65 | |||
XM_021068554.1 | XP_020924213.1 | 1264 | 65 | |||
XM_021068555.1 | XP_020924214.1 | 635 | 65 | |||
XM_021068556.1 | XP_020924215.1 | 454 | 65 | |||
XM_021068557.1 | XP_020924216.1 | 540 | 65 | |||
XM_021068558.1 | XP_020924217.1 | 3850 | 65 | |||
XM_021068559.1 | XP_020924218.1 | 1263 | 65 | |||
XM_021068560.1 | XP_020924219.1 | 1330 | 65 | |||
XM_021068561.1 | XP_020924220.1 | 537 | 65 | |||
XM_021068562.1 | XP_020924221.1 | 663 | 65 | |||
GenBank mRNA | AK236380.1 | - | 5 | 532 | 65 | |
AY253340.1 | - | 1 | 152 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
