Assay Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214371.1 | NP_999536.1 | 4 | 346 | 70 | |
| XM_005672949.3 | XP_005673006.1 | 267 | 70 | |||
| XM_005672950.3 | XP_005673007.1 | 283 | 70 | |||
| XM_021077309.1 | XP_020932968.1 | 229 | 70 | |||
| XM_021077310.1 | XP_020932969.1 | 341 | 70 | |||
| GenBank mRNA | AK234093.1 | - | 4 | 406 | 70 | |
| AK236169.1 | - | 3 | 585 | 70 | ||
| AK240354.1 | - | 3 | 251 | 70 | ||
| AK352197.1 | - | 3 | 251 | 70 | ||
| AK394053.1 | - | 3 | 240 | 70 | ||
| AK395999.1 | - | 3 | 70 | |||
| AK399898.1 | - | 3 | 230 | 70 | ||
| AY339895.1 | - | 4 | 346 | 70 | ||
More Information
Set Membership: |
|

PKIG
Genomic Map