Assay Details
| Gene Symbol: TPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214373.1 | NP_999538.1 | 3 | 253 | 65 | |
| GenBank mRNA | AK231403.1 | - | 510 | 65 | ||
| AK232109.1 | - | 3 | 267 | 65 | ||
| AK236630.1 | - | 3 | 298 | 65 | ||
| AK236713.1 | - | 3 | 270 | 65 | ||
| AK394426.1 | - | 3 | 300 | 65 | ||
| AK394993.1 | - | 3 | 279 | 65 | ||
| AK397472.1 | - | 3 | 304 | 65 | ||
| AK397966.1 | - | 3 | 269 | 65 | ||
| AK397982.1 | - | 3 | 452 | 65 | ||
| AK398783.1 | - | 809 | 65 | |||
| AK398932.1 | - | 578 | 65 | |||
| AK399254.1 | - | 3 | 268 | 65 | ||
| AK400690.1 | - | 3 | 318 | 65 | ||
| AY072784.1 | - | 3 | 253 | 65 | ||
More Information
Set Membership: |
|

TPT1
Genomic Map