Assay Details
| Gene Symbol: PTGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214385.1 | NP_999550.1 | 10 | 953 | 90 | |
| XM_021083385.1 | XP_020939044.1 | 1664 | 90 | |||
| XM_021083390.1 | XP_020939049.1 | 874 | 90 | |||
| GenBank mRNA | AK232993.1 | - | 11 | 1663 | 90 | |
| AK393027.1 | - | 10 | 1042 | 90 | ||
| AK398516.1 | - | 10 | 995 | 90 | ||
| AK398931.1 | - | 10 | 1069 | 90 | ||
| D49386.1 | - | 10 | 973 | 90 | ||
| U87622.1 | - | 10 | 952 | 90 | ||
More Information
Set Membership: |
|

PTGR1
Genomic Map