Assay Details
| Gene Symbol: XIAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097436.1 | NP_001090905.1 | 6 | 1454 | 103 | |
| XM_021079365.1 | XP_020935024.1 | 1964 | 103 | |||
| XM_021079366.1 | XP_020935025.1 | 1984 | 103 | |||
| XM_021079367.1 | XP_020935026.1 | 1716 | 103 | |||
| XM_021079368.1 | XP_020935027.1 | 1785 | 103 | |||
| GenBank mRNA | EF120983.1 | - | 6 | 1454 | 103 | |
More Information
Set Membership: |
|

XIAP
Genomic Map