Assay Details
| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097441.2 | NP_001090910.2 | 10 | 1151 | 99 | |
| XM_021093133.1 | XP_020948792.1 | 1105 | 99 | |||
| GenBank mRNA | AK347582.1 | - | 10 | 1251 | 99 | |
| AK348172.1 | - | 10 | 1249 | 99 | ||
| EU116354.1 | - | 10 | 1151 | 99 | ||
| EU116355.1 | - | 9 | 1001 | 99 | ||
| HQ222879.1 | - | 10 | 1151 | 99 | ||
| HQ222880.1 | - | 9 | 1001 | 99 | ||
More Information
Set Membership: |
|

TNFRSF1B
Genomic Map