Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097479.1 | NP_001090948.1 | 4 | 350 | 106 | |
| GenBank mRNA | AK343368.1 | - | 4 | 106 | ||
| AK399809.1 | - | 4 | 385 | 106 | ||
| AY610378.1 | - | 3 | 340 | 106 | ||
| DQ629167.1 | - | 4 | 350 | 106 | ||
More Information
Set Membership: |
|

RPL27
Genomic Map