Assay Details
Gene Symbol: CSTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097496.1 | NP_001090965.1 | 3 | 255 | 112 | |
GenBank mRNA | AK392189.1 | - | 3 | 286 | 112 | |
EF397605.1 | - | 3 | 221 | 112 | ||
EF483823.1 | - | 3 | 255 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
