Assay Details
Gene Symbol: CYB561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097522.1 | NP_001090991.1 | 3 | 350 | 86 | |
XM_005668676.3 | XP_005668733.1 | 928 | 86 | |||
XM_005668677.3 | XP_005668734.1 | 860 | 86 | |||
XM_013980833.2 | XP_013836287.1 | 388 | 86 | |||
XM_021066011.1 | XP_020921670.1 | 659 | 86 | |||
XM_021066012.1 | XP_020921671.1 | 384 | 86 | |||
GenBank mRNA | AK236037.1 | - | 4 | 473 | 86 | |
AK389390.1 | - | 4 | 431 | 86 | ||
D88158.1 | - | 3 | 350 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
