Assay Details
Gene Symbol: PPP2R5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100192.1 | NP_001093662.1 | 12 - 13 | 1549 | 137 | |
GenBank mRNA | AK233028.1 | - | 1 - 2 | 96 | 137 | |
AK345162.1 | - | 2 - 3 | 145 | 137 | ||
AK389471.1 | - | 2 - 3 | 182 | 137 | ||
AK390300.1 | - | 12 - 13 | 1584 | 137 | ||
DQ115798.1 | - | 12 - 13 | 1549 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
