Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110423.1 | NP_001103893.1 | 4 | 856 | 84 | |
| XM_021093187.1 | XP_020948846.1 | 1072 | 84 | |||
| XM_021093188.1 | XP_020948847.1 | 1364 | 84 | |||
| XM_021093189.1 | XP_020948848.1 | 1096 | 84 | |||
| XM_021093190.1 | XP_020948849.1 | 989 | 84 | |||
| XM_021093191.1 | XP_020948850.1 | 904 | 84 | |||
| XM_021093192.1 | XP_020948851.1 | 993 | 84 | |||
| XM_021093193.1 | XP_020948852.1 | 1257 | 84 | |||
| XM_021093195.1 | XP_020948854.1 | 1133 | 84 | |||
| GenBank mRNA | AK344052.1 | - | 5 | 928 | 84 | |
| AK390067.1 | - | 5 | 980 | 84 | ||
| EU165525.1 | - | 4 | 856 | 84 | ||
More Information
Set Membership: |
|

AQP4
Genomic Map