Assay Details
| Gene Symbol: GPX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115155.1 | NP_001108627.1 | 5 | 644 | 80 | |
| GenBank mRNA | AK392754.1 | - | 5 | 643 | 80 | |
| AK398746.1 | - | 2 | 182 | 80 | ||
| DV936676.1 | - | 5 | 644 | 80 | ||
More Information
Set Membership: |
|

GPX3
Genomic Map