Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122996.2 | NP_001116468.2 | 5 | 638 | 107 | |
| NM_001301406.1 | NP_001288335.1 | 6 | 694 | 107 | ||
| GenBank mRNA | CX059620.1 | - | 3 | 229 | 107 | |
| CX062850.1 | - | 5 | 648 | 107 | ||
| EU545197.1 | - | 5 | 553 | 107 | ||
More Information
Set Membership: |
|

OAZ3
Genomic Map