Assay Details
| Gene Symbol: ASPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123077.1 | NP_001116549.1 | 6 | 850 | 120 | |
| GenBank mRNA | EU442574.1 | - | 6 | 850 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ASPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123077.1 | NP_001116549.1 | 6 | 850 | 120 | |
| GenBank mRNA | EU442574.1 | - | 6 | 850 | 120 | |
Set Membership: |
|