Assay Details
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123086.1 | NP_001116558.1 | 5 | 550 | 69 | |
XM_021076684.1 | XP_020932343.1 | 667 | 69 | |||
GenBank mRNA | AK235574.1 | - | 5 | 582 | 69 | |
AK346055.1 | - | 5 | 617 | 69 | ||
AK394984.1 | - | 5 | 641 | 69 | ||
AK400784.1 | - | 5 | 620 | 69 | ||
EU404087.1 | - | 5 | 550 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
