Assay Details
| Gene Symbol: RHOG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123191.1 | NP_001116663.1 | 1 | 149 | 85 | |
| XM_021101938.1 | XP_020957597.1 | 305 | 85 | |||
| XM_021101939.1 | XP_020957598.1 | 1297 | 85 | |||
| XM_021101940.1 | XP_020957599.1 | 638 | 85 | |||
| XM_021101941.1 | XP_020957600.1 | 334 | 85 | |||
| GenBank mRNA | DQ917639.1 | - | 1 | 149 | 85 | |
More Information
Set Membership: |
|

RHOG
Genomic Map