Assay Details
| Gene Symbol: SOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123197.1 | NP_001116669.1 | 1 | 854 | 85 | |
| GenBank mRNA | AK349255.1 | - | 1 | 1093 | 85 | |
| AK390168.1 | - | 1 | 1238 | 85 | ||
| AK396800.1 | - | 1 | 1281 | 85 | ||
| DQ159208.2 | - | 1 | 649 | 85 | ||
| EU503117.1 | - | 1 | 854 | 85 | ||
| EU519824.1 | - | 1 | 854 | 85 | ||
| FJ882401.1 | - | 1 | 855 | 85 | ||
More Information
Set Membership: |
|

SOX2
Genomic Map