Assay Details
Gene Symbol: TLR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348771.1 | NP_001335700.1 | 2 | 2517 | 58 | |
GenBank mRNA | FJ668383.1 | - | 2 | 2520 | 58 | |
FJ754217.1 | - | 2 | 2520 | 58 | ||
GQ248712.1 | - | 1 | 2450 | 58 | ||
KF019633.1 | - | 1 | 2450 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
