Assay Details
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213856.2 | NP_999021.2 | 4 | 351 | 80 | |
XM_021082121.1 | XP_020937780.1 | 382 | 80 | |||
GenBank mRNA | AY177149.1 | - | 4 | 329 | 80 | |
S77587.1 | - | 4 | 351 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
