Assay Details
| Gene Symbol: LGALS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001867.1 | NP_001001867.1 | 3 - 4 | 330 | 62 | |
| GenBank mRNA | AY885253.1 | - | 3 - 4 | 266 | 62 | |
| DQ367936.1 | - | 3 - 4 | 330 | 62 | ||
More Information
Set Membership: |
|

LGALS1
Genomic Map