Assay Details
| Gene Symbol: VPREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075117.1 | NP_001068585.1 | 1 - 2 | 96 | 75 | |
| GenBank mRNA | DQ904034.1 | - | 1 - 2 | 96 | 75 | |
More Information
Set Membership: |
|

VPREB1
Genomic Map