Assay Details
| Gene Symbol: UBE2J1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077219.1 | NP_001070687.1 | 5 - 6 | 420 | 93 | |
| GenBank mRNA | AK234645.1 | - | 5 - 6 | 778 | 93 | |
| AY609701.1 | - | 5 - 6 | 542 | 93 | ||
| DQ972953.1 | - | 5 - 6 | 420 | 93 | ||
More Information
Set Membership: |
|

UBE2J1
Genomic Map