Assay Details
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077226.1 | NP_001070694.1 | 3 - 4 | 324 | 74 | |
XM_005664070.3 | XP_005664127.1 | 529 | 74 | |||
XM_013997827.2 | XP_013853281.1 | 1488 | 74 | |||
GenBank mRNA | AK348596.1 | - | 5 - 6 | 486 | 74 | |
AK396389.1 | - | 5 - 6 | 483 | 74 | ||
DQ972959.1 | - | 3 - 4 | 324 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
