Assay Details
| Gene Symbol: NAXE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078664.1 | NP_001072132.1 | 5 - 6 | 659 | 82 | |
| GenBank mRNA | AK237183.1 | - | 5 - 6 | 720 | 82 | |
| DQ826508.1 | - | 5 - 6 | 659 | 82 | ||
More Information
Set Membership: |
|

NAXE
Genomic Map