Assay Details
| Gene Symbol: ARF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078677.1 | NP_001072145.1 | 1 | 349 | 127 | |
| XM_005659939.3 | XP_005659996.1 | 1120 | 127 | |||
| XM_021084333.1 | XP_020939992.1 | 1949 | 127 | |||
| GenBank mRNA | AY609726.1 | - | 2 | 862 | 127 | |
| DQ915493.1 | - | 1 | 349 | 127 | ||
| FJ455238.1 | - | 1 | 446 | 127 | ||
More Information
Set Membership: |
|

ARF6
Genomic Map