Assay Details
| Gene Symbol: HMBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097412.1 | NP_001090881.1 | 7 - 8 | 526 | 59 | |
| XM_021102144.1 | XP_020957803.1 | 708 | 59 | |||
| XM_021102146.1 | XP_020957805.1 | 839 | 59 | |||
| XM_021102147.1 | XP_020957806.1 | 712 | 59 | |||
| XM_021102148.1 | XP_020957807.1 | 657 | 59 | |||
| GenBank mRNA | AK238444.1 | - | 7 - 8 | 560 | 59 | |
| DQ845174.1 | - | 7 - 8 | 526 | 59 | ||
More Information
Set Membership: |
|

HMBS
Genomic Map