Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097415.1 | NP_001090884.1 | 3 - 4 | 391 | 65 | |
| GenBank mRNA | AB190766.1 | - | 3 - 4 | 391 | 65 | |
| AK231170.1 | - | 3 - 4 | 383 | 65 | ||
| AK237721.1 | - | 3 - 4 | 377 | 65 | ||
| AK346556.1 | - | 3 - 4 | 363 | 65 | ||
| AK346608.1 | - | 3 - 4 | 384 | 65 | ||
| AK346874.1 | - | 3 - 4 | 370 | 65 | ||
| AK346937.1 | - | 3 - 4 | 380 | 65 | ||
| AY459537.2 | - | 3 - 4 | 323 | 65 | ||
More Information
Set Membership: |
|

LY86
Genomic Map