Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097415.1 | NP_001090884.1 | 4 - 5 | 456 | 70 | |
| GenBank mRNA | AB190766.1 | - | 4 - 5 | 456 | 70 | |
| AK231170.1 | - | 4 - 5 | 70 | |||
| AK346556.1 | - | 4 - 5 | 70 | |||
| AK346608.1 | - | 4 - 5 | 70 | |||
| AK346874.1 | - | 4 - 5 | 70 | |||
| AY459537.2 | - | 4 - 5 | 388 | 70 | ||
More Information
Set Membership: |
|

LY86
Genomic Map